Skip to Content
Merck
All Photos(1)

Key Documents

NIST2260A

Aromatic hydrocarbons in toluene

NIST® SRM® 2260a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1.2 mL

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is a solution of aromatic hydrocarbons, primarily polycyclic aromatic hydrocarbons (PAHs) ranging in molecular mass from 128u to 302u, in toluene. This SRM is intended for use in the calibration of chromatographic instrumentation used for the determination of aromatic hydrocarbons, primarily PAHs. For more information, please refer to the SDS and COA.

SRM 2260A_cert

SRM 2260A_SDS

Application


  • Determination of retinol and retinyl esters in human plasma by high-performance liquid chromatography with automated column switching and ultraviolet detection.: This research utilizes the Superspher® 100 RP-18 endcapped LICHROCART 125-4 column for the precise measurement of retinol and retinyl esters in human plasma, demonstrating high efficiency and accuracy in the HPLC method (Hartmann et al., 2001).

  • Determination of endogenous levels of 13-cis-retinoic acid (isotretinoin), all-trans-retinoic acid (tretinoin) and their 4-oxo metabolites in human and animal plasma by high-performance liquid chromatography with automated column switching and ultraviolet detection.: Utilizing the Superspher® 100 RP-18 endcapped column, this research develops a method for determining endogenous retinoic acids and their metabolites, emphasizing its application in pharmacokinetic studies (Wyss & Bucheli, 1997).

  • High-performance liquid chromatographic enantioselective assay for the measurement of ketoprofen glucuronidation by liver microsomes.: This paper discusses the enantioselective assay using the Superspher® 100 RP-18 endcapped column, demonstrating its effectiveness in the measurement of ketoprofen glucuronidation in liver microsomes (Chakir et al., 1994).

  • Rapid liquid chromatographic assay of glutathione in cultured cells.: This study highlights the rapid and efficient liquid chromatographic assay of glutathione in cultured cells using the Superspher® 100 RP-18 endcapped column, showcasing its utility in biochemical analysis (Leroy et al., 1993).



Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Naphthalene, Biphenyl, Acenaphthylene, Acenaphthene, Fluorene, Dibenzothiophene, Phenanthrene, Anthracene, 4H-Cyclopenta[def]phenanthrene, Fluoranthene, Pyrene, Benzo[ghi]fluoranthene, Cyclopenta[cd]pyrene, Benzo[c]phenanthrene, Benz[a]anthracene, Chrysene, Triphenylene, Benzo[b]fluoranthene, Benzo[j]fluoranthene, Benzo[k]fluoranthene, Benzo[a]fluoranthene, Benzo[e]pyrene, Benzo[a]pyrene, Perylene, Indeno[1,2,3-cd]pyrene, Benzo[ghi]perylene, Dibenz[a,h]anthracene, Dibenz[a,c]anthracene, Dibenz[a,j]anthracene, Picene , Benzo[b]chrysene, Anthanthrene, Coronene, Dibenzo[b,k]fluoranthene, Dibenzo[a,e]pyrene

See certificate for values and more details at nist.gov/SRM.

Legal Information

LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Superspher is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service