Skip to Content

ERMEF412

Brown coal (GCV, NCV, element content and trace elements)

ERM®, certified reference material

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

agency

ERM®

manufacturer/tradename

JRC

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

Certified for gross calorific value (GCV), net calorific value (NCV), volatile matter, fly ash, C, H, N, S, Ca, Hg, K, Mn, Na, Se, V.

Analysis Note

A JRC Certificate of Analysis is included with the product, a pdf copy of the CoA and other documentation you can find here:
please visit the JRC webpage

Legal Information

ERM is a registered trademark of European Commission


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library