Skip to Content
Merck
All Photos(1)

Key Documents

936898

Sigma-Aldrich

SPhos Pd G3 ChemBeads

Synonym(s):

(2-Dicyclohexylphosphino-2′,6′-dimethoxybiphenyl) [2-(2′-amino-1,1′-biphenyl)]palladium(II) methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H48NO5PPdS
CAS Number:
Molecular Weight:
780.26
MDL number:
UNSPSC Code:
31191513
NACRES:
NA.22

form

solid

Quality Level

composition

, ~5 wt. % (loading of catalyst)

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

InChI

InChI: 1S/C26H35O2P.C12H10N.CH4O3S.Pd/c1-27-23-17-11-18-24(28-2)26(23)22-16-9-10-19-25(22)29(20-12-5-3-6-13-20)21-14-7-4-8-15-21;13-12-9-5-4-8-11(12)10-6-2-1-3-7-10;1-5(2,3)4;/h9-11,16-21H,3-8,12-15H2,1-2H3;1-6,8-9H,13H2;1H3,(H,2,3,4);/q;;;+1/p-1

InChI key

SCWODMZBSVVMRH-UHFFFAOYSA-M

General description

SPhos Pd G3 is a third-generation (G3) Buchwald precatalyst that can be used in cross-coupling reactions for the formation of C-C, C-N, C-O, C-F, C-CF3, and C-S bonds. It is air-, moisture-, and thermally stable and is highly soluble in a wide range of common organic solvents. Some of its unique features include lower catalyst loadings, shorter reaction time, efficient formation of the active catalytic species, and accurate control of ligand: palladium ratio.

Application

Pd catalyst for cross-coupling SPhos Pd G3 can be used as a precatalyst in the Suzuki–Miyaura catalyst–transfer polymerization (SCTP) reaction of a wide spectrum of monomers, including electron-rich to electron-deficient (hetero)arenes. It is also used as a catalyst in the formation of a Csp3–Csp2 bond between sterically hindered boronic hemiester and quinone diazide, which is the key intermediate step in the enantioselective synthesis of azamerone. ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

For larger scale uses, product also available in powdered form (776246)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana L Aguirre et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(51), 12981-12986 (2021-07-08)
High-throughput experimentation (HTE) methods are central to modern medicinal chemistry. While many HTE approaches to C-N and Csp2 -Csp2 bonds are available, options for Csp2 -Csp3 bonds are limited. We report here how the adaptation of nickel-catalyzed cross-electrophile coupling of
Noah P Tu et al.
Angewandte Chemie (International ed. in English), 58(24), 7987-7991 (2019-03-21)
Technologies that enable rapid screening of diverse reaction conditions are of critical importance to methodology development and reaction optimization, especially when molecules of high complexity and scarcity are involved. The lack of a general solid dispensing method for chemical reagents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service