Skip to Content
Merck
All Photos(1)

Key Documents

90257

Supelco

2,3,4,5,6-Pentafluorobenzyl bromide

for GC derivatization, LiChropur, ≥98.5% (GC)

Synonym(s):

PFB-Br, α-Bromo-2,3,4,5,6-pentafluorotoluene, (Bromomethyl)pentafluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5CH2Br
CAS Number:
Molecular Weight:
260.99
Beilstein:
647808
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.05

grade

for GC derivatization

Quality Level

Assay

≥98.5% (GC)

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.471 (lit.)

bp

174-175 °C (lit.)

mp

19-20 °C (lit.)

density

1.728 g/mL at 25 °C (lit.)

SMILES string

Fc1c(F)c(F)c(CBr)c(F)c1F

InChI

1S/C7H2BrF5/c8-1-2-3(9)5(11)7(13)6(12)4(2)10/h1H2

InChI key

XDEPVFFKOVDUNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Learn more in the Product Information
It is used for the analysis of D2/E2-isoprostanes using negative ion chemical ionization-gas chromatography/mass spectrometry (NICI-GC/MS). Fatty acids may be converted to pentafluorobenzyl esters using PFG-Br and diisopropylethylamine during determination of fatty acids using NICI-GC/MS.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service