Skip to Content
Merck
All Photos(1)

Key Documents

57618

Supelco

Potassium phosphate monobasic

suitable for HPLC, LiChropur, 99.5-101.0% (T)

Synonym(s):

Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
EC Number:
MDL number:
UNSPSC Code:
41116105
NACRES:
NB.21
Pricing and availability is not currently available.

Assay

99.5-101.0% (T)

form

solid

quality

LiChropur

technique(s)

HPLC: suitable

pH

4.1-4.5

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

252.6 °C (lit.)

solubility

H2O: 10%, clear, colorless

density

2.338 g/mL at 25 °C (lit.)

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: ≤0.1
λ: 220 nm Amax: ≤0.06
λ: 230 nm Amax: ≤0.04
λ: 300 nm Amax: ≤0.02
λ: 500 nm Amax: ≤0.02

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Partitioning recombinant chitinase from Nicotiana benthamiana by an aqueous two-phase system: Demonstrates the application of potassium phosphate monobasic in aqueous two-phase systems for efficient partitioning of enzymes, which is crucial for biotechnological and industrial applications (Tue et al., 2024).

  • Unlocking Nature′s Toolbox: This study showcases how potassium phosphate monobasic can be used to optimize conditions for the production of recombinant proteins, enhancing yields and streamlining production processes in biomanufacturing (Rajak et al., 2024).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service