Skip to Content
Merck
All Photos(1)

Documents

57522

Supelco

Isoeugenol, predominantly trans

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (sum of cis- and trans-isomers, GC)

shelf life

limited shelf life, expiry date on the label

concentration

≤5.0% (cis-isomer, GC)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

InChI

1S/C10H12O2/c1-3-4-8-5-6-9(11)10(7-8)12-2/h3-7,11H,1-2H3/b4-3+

InChI key

BJIOGJUNALELMI-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isoeugenol belongs to the class of phenylpropenes, a naturally occurring aroma compound found in various plant species. It is predominantly used as a flavoring agent in food products and beverages. Owing to its antioxidant and anti-inflammatory activities, it is commonly used in cosmetics, dentistry, and traditional medicine.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service