Skip to Content
Merck

41519

RBS™ A 285 SOLID pF

≥20.0% Na, powder, low foam, phosphate-free

Synonym(s):

Phosphate free alkaline detergent

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41103206
NACRES:
NA.22
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

RBS™ A 285 SOLID pF, low foam, phosphate-free

Quality Segment

form

powder

concentration

≥20.0% Na

pH

11.0-12.0(at 2% v/v in water)

General description

RBS™ A 285 SOLID pF is a highly efficient powdered phosphate free alkaline detergent specifically formulated for use in laboratory washing machines.

Application

RBS™ A 285 SOLID pF is recommended for cleaning various laboratory glassware and equipment, including those made of stainless steel, plastic, and porcelain. The detergent is designed to effectively remove distillation residues, proteins, oils, and greases.

Features and Benefits

  • Save money by using less detergent because it’s efficient at low concentrations
  • Avoid contamination by thoroughly removing residues and contaminants with strong cleaning and degreasing power
  • Gain confidence in your clean with reliable outcomes no matter the water hardness
  • Reach your sustainability goals with an environmentally friendly product that’s 100% biodegradable

Legal Information

RBS is a trademark of Chemical Products R. Borghgraef S.A.


Pictograms

Corrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library