Skip to Content
Merck
All Photos(2)

Documents

34083

Supelco

Flunixin-d3

VETRANAL®, analytical standard

Synonym(s):

2-[2-Methyl-d3-3-(trifluoromethyl)phenylamino]nicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8D3F3N2O2
CAS Number:
Molecular Weight:
299.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])c1c(Nc2ncccc2C(O)=O)cccc1C(F)(F)F

InChI

1S/C14H11F3N2O2/c1-8-10(14(15,16)17)5-2-6-11(8)19-12-9(13(20)21)4-3-7-18-12/h2-7H,1H3,(H,18,19)(H,20,21)/i1D3

InChI key

NOOCSNJCXJYGPE-FIBGUPNXSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Abir Khaled et al.
Analytica chimica acta, 1056, 34-46 (2019-02-25)
This paper presents the development and validation of a fully automated, high-throughput multiclass, multiresidue method for quantitative analysis of 77 veterinary drugs in chicken muscle via direct immersion solid phase microextraction (DI-SPME) and ultra-high pressure liquid chromatography-electrospray ionization - tandem
Weilin L Shelver et al.
Journal of agricultural and food chemistry, 64(1), 336-345 (2015-12-24)
Twenty cull dairy cows (645 ± 83 kg) were treated with 2.2 mg/kg bw flunixin by intravenous (IV) or intramuscular (IM) administration with, or without, exposure to lipopolysaccharide in a two factor balanced design. The usefulness of screening assays to
Weilin L Shelver et al.
Journal of agricultural and food chemistry, 64(51), 9697-9701 (2016-12-14)
Twenty dairy cows received flunixin meglumine at 2.2 mg/kg bw, administered once daily by either the intravenous (IV) or intramuscular (IM) route for three consecutive days with either intravenous normal saline (NS) or lipopolysaccharide (LPS) providing a balanced design with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service