Skip to Content
Merck
All Photos(1)

Documents

16587

Supelco

Daidzein

analytical standard

Synonym(s):

4′,7-Dihydroxy­iso­flavone, 7-Hydroxy-3-(4-hydroxy­phenyl)-4H-1-benzo­pyran-4-one, 7-Hydroxy-3-(4-hydroxy­phenyl)­chromone, Isoaurostatin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O4
CAS Number:
Molecular Weight:
254.24
Beilstein:
231523
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1)C2=COc3cc(O)ccc3C2=O

InChI

1S/C15H10O4/c16-10-3-1-9(2-4-10)13-8-19-14-7-11(17)5-6-12(14)15(13)18/h1-8,16-17H

InChI key

ZQSIJRDFPHDXIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Daidzein is an endocrine-active estrogenic isoflavone. This phytoestrogen is capable to bind to estrogen to exert estrogenic effects in vivo. It can naturally be found in soybean seed and flour.

Application

Daidzein was used as standard in determination of daidzein and genistein, in soybean food using in-tube solid-phase microextraction coupled to high-performance liquid chromatography (in-tube SPME–HPLC). It was also used as reference standard in determination of isoflavones in red clover using high-performance liquid chromatography–UV–electrospray ionization-mass spectrometric detector (HPLC–UV–ESI-MSD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Soy isoflavone daidzein protects against oxidative damage in liver cells induced by 7,12-dimethylbenz[a]anthracene (DMBA). Catalase and superoxide dismutase activity, down-regulated by DMBA, was restored by daidzein.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of daidzein and genistein in soybean foods by automated on-line in-tube solid-phase microextraction coupled to high-performance liquid chromatography.
Mitani K, Narimatsu S, Kataoka H.
Journal of Chromatography A, 986(2), 169-177 (2003)
Determination of isoflavones in red clover and related species by high-performance liquid chromatography combined with ultraviolet and mass spectrometric detection.
Wu Q, Wang M, Simon JE.
Journal of Chromatography A, 1016(2), 195-209 (2003)
Developmental effects of dietary phytoestrogens in Sprague-Dawley rats and interactions of genistein and daidzein with rat estrogen receptors alpha and beta in vitro.
Casanova, Mercedes, et al.
Toxicological Sciences, 51.2, 236-244 (1999)
Ivonne M C M Rietjens et al.
Molecular nutrition & food research, 57(1), 100-113 (2012-11-24)
Isoflavones are phytoestrogens that have been linked to both beneficial as well as adverse effects in relation to cell proliferation and cancer risks. The present article presents an overview of these seemingly contradicting health effects and of mechanisms that could
Vera van der Velpen et al.
The Journal of nutrition, 143(6), 774-780 (2013-04-26)
Isoflavones (genistein, daidzein, and glycitein) are suggested to have benefits as well as risks for human health. Approximately one-third of the Western population is able to metabolize daidzein into the more potent metabolite equol. Having little endogenous estradiol, equol-producing postmenopausal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service