Skip to Content
Merck
All Photos(1)

Documents

05-5150

Sigma-Aldrich

Cobalt(II) chloride hexahydrate

JIS special grade, ≥99.0%

Synonym(s):

Cobaltous chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoCl2 · 6H2O
CAS Number:
Molecular Weight:
237.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

vapor pressure

40 mmHg ( 0 °C)

Assay

≥99.0%

form

solid

reaction suitability

reagent type: catalyst
core: cobalt

availability

available only in Japan

SMILES string

O.O.O.O.O.O.Cl[Co]Cl

InChI

1S/2ClH.Co.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

GFHNAMRJFCEERV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Dam. 1 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asma Tabasum et al.
Plants (Basel, Switzerland), 10(1) (2020-12-31)
Pesticides are one of the main organic pollutants as they are highly toxic and extensively used worldwide. The reclamation of wastewater containing pesticides is of utmost importance. For this purpose, GO-doped metal ferrites (GO-Fe3O4 and GO-CoFe2O4) were prepared and characterized
H Lawrence et al.
The bone & joint journal, 96-B(9), 1172-1177 (2014-09-04)
Abnormal wear of cobalt-containing metal-on-metal joints is associated with inflammatory pseudotumours. Cobalt ions activate human toll-like receptor 4 (TLR4), which normally responds to bacterial lipopolysaccharide (LPS) in sepsis. Activation of TLR4 by LPS increases the expression of chemokines IL-8 and
Junya Nagai et al.
Biochemical and biophysical research communications, 450(1), 476-481 (2014-06-14)
Recently, we found that albumin overload induces expression of the transcription factor hypoxia-inducible factor-1α (HIF-1α) protein and several HIF-1 target genes in human renal proximal tubular epithelial cell line HK-2. In this study, the role of albumin-bound fatty acids in
Gláucia Melo Reis et al.
Pharmacology, biochemistry, and behavior, 131, 112-118 (2015-02-18)
The stimulation of the retrosplenial cortex (RSC) is antinociceptive in the rat tail-flick and formalin tests. The rat RSC is caudal to and send projections to the ipsilateral anterior cingulate cortex (ACC), which is also involved in pain processing. This
Juhui Ko et al.
Journal of hazardous materials, 285, 11-17 (2014-12-03)
Aflatoxin B1 (AFB1) is a well-known carcinogenic contaminant in foods. It is classified as an extremely hazardous compound because of its potential toxicity to the human nervous system. AFB1 has also been extensively used as a biochemical marker to evaluate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service