Skip to Content
Merck
All Photos(1)

Key Documents

05-2650

Sigma-Aldrich

Chloral hydrate

SAJ special grade, ≥99.7%

Synonym(s):

Trichloroacetaldehyde hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCH(OH)2
CAS Number:
Molecular Weight:
165.40
Beilstein:
1698497
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

SAJ special grade

Assay

≥99.7%

availability

available only in Japan

mp

57 °C (lit.)

SMILES string

OC(O)C(Cl)(Cl)Cl

InChI

1S/C2H3Cl3O2/c3-2(4,5)1(6)7/h1,6-7H

InChI key

RNFNDJAIBTYOQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie K West et al.
The British journal of ophthalmology, 97(11), 1437-1442 (2013-09-21)
To report the largest study on the safety and effectiveness of sedation in paediatric ophthalmology in a nurse-led outpatient sedation unit. Retrospective cohort study reviewing all children who underwent sedation from January 2006 to December 2010. Patients were sedated with
Tal I Arnon et al.
PloS one, 3(5), e2150-e2150 (2008-05-15)
The natural cytotoxic receptors (NCRs) are a unique set of activating proteins expressed mainly on the surface of natural killer (NK) cells. The NCRs, which include three members; NKp46, NKp44 and NKp30, are critically involved in NK cytotoxicity against different
A D Steinberg
Pediatrics, 92(3), 442-446 (1993-09-01)
Current federal regulations of potentially carcinogenic environmental chemicals are based on the assumption that risks for humans can be extrapolated from the effects of chronic high-dose exposure of rodents. It is assumed that all chemicals induce cancer by a genotoxic
Nicholas M Allen et al.
Developmental medicine and child neurology, 56(2), 105-112 (2013-12-07)
Status dystonicus is a rare, but life-threatening movement disorder emergency. Urgent assessment is required and management is tailored to patient characteristics and complications. The use of dystonia action plans and early recognition of worsening dystonia may potentially facilitate intervention or
Simone Jürschik et al.
Journal of mass spectrometry : JMS, 47(9), 1092-1097 (2012-09-14)
In this work, we illustrate the application of proton transfer reaction mass spectrometry (PTR-MS) in the field of food and drink safety. We present proof-of-principle measurements of four different drinks (water, tea, red wine and white wine) each spiked separately

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service