Skip to Content
Merck
All Photos(1)

Key Documents

05-2510

Sigma-Aldrich

Cetyl alcohol

SAJ special grade, ≥98.0%

Synonym(s):

1-Hexadecanol, Palmityl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15OH
CAS Number:
Molecular Weight:
242.44
Beilstein:
1748475
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

SAJ special grade

Assay

≥98.0%

form

solid

availability

available only in Japan

bp

190 °C/15 mmHg (lit.)

SMILES string

CCCCCCCCCCCCCCCCO

InChI

1S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3

InChI key

BXWNKGSJHAJOGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toru Uyama et al.
The Journal of biological chemistry, 290(28), 17520-17534 (2015-05-29)
Phospholipase A/acyltransferase (PLA/AT)-3 (also known as H-rev107 or AdPLA) was originally isolated as a tumor suppressor and was later shown to have phospholipase A1/A2 activity. We have also found that the overexpression of PLA/AT-3 in mammalian cells results in specific
N Dashti et al.
Chemosphere, 70(3), 475-479 (2007-08-07)
Bacteria and fungi in pristine and oily desert soil samples were counted on inorganic medium aliquots containing 0.5% hexadecane, hexadecanol, hexadecanal or hexadecanoic acid, as sole sources of carbon and energy. It was found that the carbon and energy source
Maria K Ratajczak et al.
Biophysical journal, 93(6), 2038-2047 (2007-05-29)
Adding cholesterol to monolayers of certain phospholipids drives the separation of liquid-ordered from liquid-disordered domains. The ordered phases appear to contain stoichiometric complexes of cholesterol and phospholipid. Furthermore, it has been suggested that the cholesterol in these complexes has a
Dhanalakshmi S Anbukumar et al.
Journal of lipid research, 51(5), 1085-1092 (2009-12-19)
Neutrophils are important in the host response against invading pathogens. One chemical defense mechanism employed by neutrophils involves the production of myeloperoxidase (MPO)-derived HOCl. 2-Chlorohexadecanal (2-ClHDA) is a naturally occurring lipid product of HOCl targeting the vinyl ether bond of
Luca Lo Piccolo et al.
Applied and environmental microbiology, 77(4), 1204-1213 (2010-12-25)
Enzymes involved in oxidation of long-chain n-alkanes are still not well known, especially those in gram-positive bacteria. This work describes the alkane degradation system of the n-alkane degrader actinobacterium Gordonia sp. strain SoCg, which is able to grow on n-alkanes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service