Skip to Content
Merck

TX1060

1,2,4-Trichlorobenzene

Meets ACS Specifications

Synonym(s):

1,2,4-Trichlorobenzene, acs solvents

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
1,2,4-C6H3Cl3
CAS Number:
UNSPSC Code:
12352101
EC Index Number:
204-428-0
MDL number:
Assay:
≥99% (corrected for water content, GC)
Grade:
ACS reagent
Bp:
213.5 °C/1013 hPa
Vapor pressure:
1.3 hPa ( 20 °C)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

vapor pressure

1.3 hPa ( 20 °C)

assay

≥99% (corrected for water content, GC)

form

liquid

dilution

(for analytical testing)

impurities

≤0.0003 meq/g Titrable acid, ≤0.02% Water (H2O)

color

APHA: ≤10

bp

213.5 °C/1013 hPa

mp

17 °C

transition temp

flash point 99 °C

solubility

0.049 g/L

density

1.45 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

Clc1c(ccc(c1)Cl)Cl

InChI

1S/C6H3Cl3/c7-4-1-2-5(8)6(9)3-4/h1-3H

InChI key

PBKONEOXTCPAFI-UHFFFAOYSA-N

Application

Meets ACS Specifications

Analysis Note

Assay (GC, corrected for water content)99.0% minColor (APHA)10 maxFormClear liquidIdentity (IR-spectrum)ConformsTitrable acid0.0003meq/g maxWater (H2O)0.02% max


Still not finding the right product?

Explore all of our products under 1,2,4-Trichlorobenzene


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library