Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
EC Index Number:
203-625-9
UNSPSC Code:
12352005
Beilstein/REAXYS Number:
635760
MDL number:
Assay:
≥98% (GC)
Bp:
110-111 °C (lit.)
Vapor pressure:
22 mmHg ( 20 °C), 26 mmHg ( 25 °C)
vapor density
3.2 (vs air)
Quality Segment
vapor pressure
22 mmHg ( 20 °C), 26 mmHg ( 25 °C)
assay
≥98% (GC)
form
liquid
autoignition temp.
997 °F
expl. lim.
7 %
color
APHA: ≤40
refractive index
n
bp
110-111 °C (lit.)
mp
-93 °C (lit.)
transition temp
flash point 4 °C
solubility
0.52 g/L
density
0.865 g/mL at 25 °C (lit.)
application(s)
histology
shipped in
ambient
storage temp.
room temp
SMILES string
Cc1ccccc1
InChI
1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3
InChI key
YXFVVABEGXRONW-UHFFFAOYSA-N
General description
EMPARTA solvents are designed for routine chemical synthesis, drying, purification, and critical labware cleaning in quality control environments. Engineered for precision, they deliver consistently reproducible results essential for rigorous validation and documentation.
Exceptional Quality Standards: These solvents meet or exceed ACS specifications, providing assurance of high quality that is crucial for accurate testing and analysis in quality control.
Consistent Reproducibility: Designed for precision, the solvents ensure that results can be reliably reproduced, which is vital for maintaining compliance and integrity in quality control processes.
Versatile Applications: Suitable for a variety of QC tasks, including analytical testing, method validation, and testing USP/NF monographs, these solvents support a comprehensive approach to quality assurance in laboratory settings.
Exceptional Quality Standards: These solvents meet or exceed ACS specifications, providing assurance of high quality that is crucial for accurate testing and analysis in quality control.
Consistent Reproducibility: Designed for precision, the solvents ensure that results can be reliably reproduced, which is vital for maintaining compliance and integrity in quality control processes.
Versatile Applications: Suitable for a variety of QC tasks, including analytical testing, method validation, and testing USP/NF monographs, these solvents support a comprehensive approach to quality assurance in laboratory settings.
Analysis Note
Assay (GC): 98% min
Color (APHA): 40 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Color (APHA): 40 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Still not finding the right product?
Explore all of our products under Toluene
signalword
Danger
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 3
flash_point_f
39.9 °F - closed cup
flash_point_c
4.4 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


