Skip to Content
Merck

TX0700

Titanium Trichloride Solution

20% w/w, technical grade

Synonym(s):

Titanium Trichloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Titanium Trichloride Solution 20% W/W, technical grade

grade

technical grade

Quality Segment

form

liquid

shipped in

ambient

storage temp.

room temp

General description

Technical

Application

Technical

Analysis Note

Assay: ~20% min
Color: Violet
Form: Liquid


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library