Skip to Content
Merck
All Photos(1)

Key Documents

SX0390

Supelco

Sodium Bromide

Meets ACS Specifications GR ACS

Synonym(s):

Sodium Bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BrNa
CAS Number:
Molecular Weight:
102.89
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-599-9
grade:
ACS reagent
form:
crystals
solubility:
946 g/L

grade

ACS reagent

Quality Level

vapor pressure

1.3 hPa ( 806 °C)

Assay

≥99.0%

form

crystals

impurities

≤0.005% Insoluble matter

pH

5.74 (22.5 °C, 430 g/L in H2O)

solubility

946 g/L

anion traces

bromate (BrO3-): ≤0.001%
chloride (Cl-): ≤0.2%
sulfate (SO42-): ≤0.002%

cation traces

Ba: ≤0.002%
Ca: ≤0.002%
Fe: ≤5 ppm
K: ≤0.1%
Mg: ≤0.001%
heavy metals (as Pb): ≤5 ppm

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].[Br-]

InChI

1S/BrH.Na/h1H;/q;+1/p-1

InChI key

JHJLBTNAGRQEKS-UHFFFAOYSA-M

Related Categories

Application

Meets ACS Specifications GR ACS

Analysis Note

Assay (corrected)99.0% minBarium (Ba)0.002% maxBromate (BrO3)0.001% maxCalcium (Ca)0.002% maxChloride (Cl)0.2% maxHeavy metals (as Pb)5ppm maxInsoluble matter0.005% maxIron (Fe)5ppm maxMagnesium (Mg)0.001% maxPotassium (K)0.1% maxSulfate (SO4)0.002% maxpH of a 5% solution at 25°C5.0-8.8

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service