QPAK00TEX
QPAK® Purification Cartridge
Designed for use with Milli-Q® Direct systems for removal of organic and ionic contaminants down to trace levels, when fed with purified water
Synonym(s):
QPAK® TEX Polishing Cartridge
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.14
Recommended Products
Quality Level
packaging
pkg of 1 unit
manufacturer/tradename
QPAK®
resistivity
>18.0 MΩ-cm (at 25 degree celcius)
technique(s)
ion chromatography: suitable
total organic carbon (TOC) residue
<10 ppb
compatibility
for use with Milli-Q® Direct
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
QPAK® TEX Polishing Cartridge is designed to remove ionic and organic contaminants down to trace levels, when fed with purified water. The cartridge is constructed with clean and high-purity materials to minimize leaching of extractable contaminants. For use with Milli-Q® Direct systems.
Application
Removes ionic and organic contaminants down to trace levels.
Features and Benefits
- Genuine consumable for the replacement of QPAK® – QPAK00TEX cartridge filter.
- e-Sure radio-frequency identification (RFID) tag ensures full traceability and provides instant visibility on pack parameters.
- Uses organex and ion-exchange resins.
- Certificate of Quality available online.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
QPAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service