Skip to Content

PP540-M

Mouse IgG Purified (1mg)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32160702
UNSPSC Code:
12352203
Species reactivity:
mouse
Species reactivity:
-
Application:
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody form

purified antibody

Quality Segment

species reactivity

mouse

Preparation Note

Refrigerator (2–8°C)

Disclaimer

This product is intended for use by IVD manufacturers.
Contact customer service for assistance: oem.reagents@emdmillipore.com
Minimum order quantity required to qualify. Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library