NY1H00010
Nylon Membrane Filter, 100.0 μm Pore Size
Millipore, 30 cm x 3 m, hydrophilic
Synonym(s):
Nylon Net Filter
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
Recommended Products
material
Nylon membrane
plain filter
white filter
sterility
non-sterile
feature
hydrophilic
manufacturer/tradename
Millipore
parameter
100 °C max. temp. (curling may be seen at temperatures above 75°C)
filter L × W
30 cm × 3 m
thickness
80 μm
pore size
100.0 μm pore size
44 % porosity
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
General description
Filter Code: NY1H
Filter Type: Net filter
Nylon filters are compatible with a broad range of solvents. Two types are available: 1) Nylon Membrane Filters with pore sizes ranging from 0.20 to 1.2 µm, and 2) Woven Net Filters with mesh openings ranging from 5 to 180 µm.
Our 5 µm Nylon Woven Net Filter (NY054700) is commonly used in the automotive industry for cleanliness analysis.
Features & Benefits:
•Wide range of pore sizes available
•Resistant to aggressive solvents
Applications:
Particle Removal and Clarification; Solvent Filtration; Particle Analysis; Paint Monitoring; Automotive Cleanliness Analysis
Our 5 µm Nylon Woven Net Filter (NY054700) is commonly used in the automotive industry for cleanliness analysis.
Features & Benefits:
•Wide range of pore sizes available
•Resistant to aggressive solvents
Applications:
Particle Removal and Clarification; Solvent Filtration; Particle Analysis; Paint Monitoring; Automotive Cleanliness Analysis
Features and Benefits
- Nylon filters display resistance to aggressive solvents.
- Compatible with a broad range of solvents.
- Applicable for use in solvent filtration, particle analysis, and clarification.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Yunlong Lu et al.
Frontiers in plant science, 6, 391-391 (2015-06-18)
The development of sperm cells (SCs) from microspores involves a set of finely regulated molecular and cellular events and the coordination of these events. The mechanisms underlying these events and their interconnections remain a major challenge. Systems analysis of genome-wide
Can Li et al.
STAR protocols, 1(3), 100195-100195 (2020-12-31)
Assessing IL-2-induced phosopho-STAT5 (pSTAT5) content can reveal the cytokine responsiveness of individual T cells. Identifying distinct T cell subsets by nuclear transcription factors, such as Foxp3, and concurrently quantifying intracellular pSTAT5, however, has been technically challenging. Conventional Foxp3 staining buffers quench pSTAT5
Artyom A Alekseyenko et al.
Current protocols in molecular biology, 109, 21-21 (2015-01-07)
In order to understand how chromatin complexes function in the nucleus, it is important to obtain a comprehensive picture of their protein, DNA, and RNA components, as well as their mutual interactions. This unit presents a chromatin cross-linking approach (BioTAP-XL)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service