Skip to Content
Merck

MSSACCEPT0R-M

96 well Collection Plate

Synonym(s):

96-Well Collection Plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41104923
NACRES:
NB.13
eCl@ss:
32039006
Material:
PTFE plate (PTFE), white plate
Sterility:
non-sterile
Manufacturer/tradename:
MultiScreen®
Packaging:
pkg of 1 collection plate
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

PTFE plate (PTFE), white plate

sterility

non-sterile

usage

Manual

packaging

pkg of 1 collection plate

manufacturer/tradename

MultiScreen®

plate size

96 wells

compatibility

manifold only

General description

Increasing drug candidate predictability earlier in the discovery pipeline using non-cell-based assays can improve candidate throughput during the ADME stage. Non-cell-based permeability assays are predictive models for oral absorption of development candidates.

MultiScreen Permeability, MultiScreen-IP, and MultiScreen Acceptor plates support artificial membrane strategies for predicting passive transport.

Membranes have uniform pore structure and density for reliable transport:

- Individual wells minimize candidate cross-talk

- Compatibility with automated liquid handling systems increases throughput

Permeability Assay:

- The MultiScreen Permeability plate has a polycarbonate membrane, which supports a hexane/hexadecane artificial layer for permeability assays.1

MultiScreen Acceptor Plate:

- The MultiScreen Acceptor plate can be used with either plate to capture passively transported compounds. Constructed of 100% PTFE, the plate can improve LC/MS results depending on the charge of the small molecules being evaluated.

Application

Non-Sterile, reusable, sturdy device made of PTFE, 1 plate
Research Category
Nucleic acid|Protein

Physical form

Color Code:Not Applicable

Other Notes

Automation Compatible:Yes

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under 96 well Collection Plate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library