Skip to Content
Merck

MP0DSYS2B

Process Scale Holders

for use with Pod Filters, 316L stainless steel

Synonym(s):

Process Scale Holders 2 Racks

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32031690
UNSPSC Code:
23151806
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

316L stainless steel , silicone seal

description

2-rack holder; for Pod configurations of 5.5 m2 to 22 m2; ITT valves

fitting

1.5 in. gauge port sanitary flange, 1.5 in. inlet/outlet sanitary flange

compatibility

for use with Pod Filters

Application

2-rack holder; for Pod configurations of 5.5 m2 to 22 m2; ITT valves


Still not finding the right product?

Explore all of our products under Process Scale Holders




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library