Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41104923
NACRES:
NB.22
eCl@ss:
32039006
Usage:
Manual
Storage temp.:
room temp
Shipped in:
ambient
sterility
non-sterile
usage
Manual
packaging
pkg of 1 ea
manufacturer/tradename
MultiScreen®
application(s)
sample preparation
shipped in
ambient
storage temp.
room temp
Other Notes
includes multiple punch and 2 carrier racks for 7 mL vials
Legal Information
MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.