Skip to Content
Merck
All Photos(3)

Key Documents

MABN2396M

Sigma-Aldrich

Anti-GCAP1 Antibody, clone 6B12

clone 6B12, from mouse

Synonym(s):

Guanylyl cyclase-activating protein 1, Guanylate cyclase activator 1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

6B12, monoclonal

species reactivity

porcine, bovine, human, rat, mouse

packaging

antibody small pack of 25 μg

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Guca1A(14913)

General description

Guanylyl cyclase-activating protein 1 (UniProt: P43081; also known as GCAP 1, Guanylate cyclase activator 1A) is encoded by the Guca1a (also known as Gcap, Gcap1, Guca1) gene (Gene ID: 14913) in murine species. GCAP1 is a membrane bound protein that stimulates retinal guanylyl cyclase when free calcium ions concentration is low and inhibits guanylyl cyclase when free calcium ions concentration is elevated. This Ca2+-sensitive regulation of retinal guanylyl cyclase appears to be a key event in recovery of the dark state of rod photoreceptors following light exposure. GCAP1 is present in rod and cone photoreceptor outer segments where phototransduction occurs. It binds three calcium ions (via EF-hands 2, 3 and 4) when calcium levels are high. Binds Mg2+ when calcium levels are low. Three calcium-binding sites are identified (aa 64-75; 100-111; and 144-155). GCAP1 restores the calcium sensitivity of guanylyl cyclase in a reconstituted system, and it decreases the sensitivity, time-to-peak, and recovery time of the light response following its introduction into intact rod outer segments. (Ref.: Gorczyca WA (1995). J. Biol Chem. 270(37); 22029-36; Schrem A et al. (1999). J Biol Chem. 274(10); 6244-49).

Specificity

Clone 6B12 detects Guanylyl cyclase-activating protein 1 in Bovine, Human, Mouse, Porcine, and Rat. It targets an epitope within 45 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
GST-fusion protein containing 45 amino acids from the C-terminal region of murine GCAP1. It does not display any homology with GCAP2.

Application

Anti-GCAP1, clone 6B12, Cat. No. MABN2396, is a highly specific mouse monoclonal antibody that targets Guanylyl cyclase-activating protein 1 and has been tested in Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
Immunofluorescence Analysis: A representative lot detected GCAP1 in Immunofluorescence applications (Zulliger, R., et. al. (2015). J Biol Chem. 290(6):3488-99).

Immunofluorescence Analysis: A 1:10 dilution from a representative lot detected GCAP1 in mouse retina.

Western Blotting Analysis: A representative lot detected GCAP1 in Western Blotting applications (Zulliger, R., et. al. (2015). J Biol Chem. 290(6):3488-99).

Immunoprecipitation Analysis: A representative lot detected GCAP1 in Immunoprecipitation applications (Zulliger, R., et. al. (2015). J Biol Chem. 290(6):3488-99).

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected GCAP1 in human retina tissue.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in C57BL/6 mouse retinal tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected GCAP1 in C57BL/6 mouse retinal tissue lysate.

Target description

23 kDa observed; 22.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service