Skip to Content
Merck
All Photos(1)

Documents

MABF2790

Sigma-Aldrich

Anti-CD96 Antibody, clone 6A6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

6A6, monoclonal

mol wt

calculated mol wt 67.13 kDa

purified by

using protein G

species reactivity

mouse

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
blocking: suitable
flow cytometry: suitable
inhibition assay: suitable

isotype

IgG2a

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Cd96(84544)

General description

T-cell surface protein tactile (UniProt: Q3U0X8; also known as Cell surface antigen CD96, T cell-activated increased late expression protein, CD96) is encoded by the Cd96 gene (Gene ID: 85444) in murine species. CD96 is a single-pass type I, disulfide-linked, homodimeric, membrane glycoprotein protein of the poliovirus receptor (PVR, CD155)-nectin family that is synthesized with a signal peptide (aa 1-21), which is subsequently cleaved off to produce the mature form that contains an extracellular domain (aa 22-536), a transmembrane domain (aa 537-557), and a cytoplasmic domain (aa 558-602). It contains two Ig-like V-type 2 domains (aa 24-134 and 138-244) and one Ig-like C2-type domain (aa 250-355). These three Ig-like domains are separated from the transmembrane domain by a long region that is rich in proline, serine, and threonine. The cytoplasmic region contains a proline-rich tandem that is flanked by arginine and lysine residues and represents binding site for SH3 domain containing signaling components. CD96 is expressed mainly by cells of hematopoietic origin, in particular on T and NK cells. It is involved in adhesive interactions of activated T and NK cells during the late phase of the immune response. It promotes NK cell-target adhesion by interacting with CD155 (PVR) present on target cells. Clone 6A6 is reported to bind to the first Ig domain of CD96 and competes with CD155 binding. It is shown to significantly reduce experimental lung metastases in murine models at low doses. (Ref.: Aguilera, AR., et al. (2018). OncoImmunology 7(5); e1424677; Georgiev, H., et al. (2018). Front. Immunol. 9; 1072; Meyer, D., et al. (2009). J. Biol. Chem. 284(4); 2235-2244).

Specificity

Clone 6A6 is a rat monoclonal antibody that detects murine CD96. It targets an epitope within the extracellular domain

Immunogen

Purified recombinant Murine CD96.

Application

Quality Control Testing

Evaluated by Flow Cytometry in EL4 cells.

Flow Cytometry Analysis: 1 μg of this antibody detected CD96 in one million EL4 cells.

Tested Applications

Flow Cytometry Analysis: A representative lot detected CD96 in Flow Cytometry applications (Seth, S., et al. (2007). Biochem Biophys Res Commun. 364(4):959-65; Aguilera, A.R., et al. (2018). Oncoimmunology. 7(5):e1424677; Chiang, E.Y., et al. (2020). Eur J Immunol. 50(6):891-902).

ELISA Analysis: A representative lot detected CD96 in ELISA applications (Aguilera, A.R., et al. (2018). Oncoimmunology. 7(5):e1424677) .

Inhibition Assay: A representative lot of this antibody blocked CD96-CD155 interaction. (Seth, S., et al. (2007). Biochem Biophys Res Commun. 364(4):959-65; Aguilera, A.R., et al. (2018). Oncoimmunology. 7(5):e1424677; Georgiev, H., et al. (2018). Front Immunol. 9:1072).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-CD96, clone 6A6, Cat. No. MABF2790, is a rat monoclonal antibody that detects CD96 (TACTILE) and is tested for use in ELISA, Flow Cytometry, Inhibition, Assay.

Physical form

Purified rat monoclonal antibody IgG2a in PBS without preservatives.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service