Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
1.3A2, monoclonal
Application:
ELISA, Neutral, WB
Citations:
-
conjugate
unconjugated
antibody form
purified antibody
clone
1.3A2, monoclonal
purified by
using protein G
species reactivity
virus, mouse
concentration
(Please refer to lot specific datasheet.)
technique(s)
ELISA: suitable, neutralization: suitable, western blot: suitable
isotype
IgG2aκ
UniProt accession no.
target post-translational modification
unmodified
General description
Sucrose gradient-purified, gamma-irradiated CHIKVMAU E2 protein (Mauritius strain 06113879) with Quil-A saponin as adjuvant followed by a second application and later a boost with binary-ethyleneimine inactivated CHIKVASIAN.
~50 kDa observed. Uncharacterized bands may be observed in some lysate(s).
Application
Western Blotting Analysis: A 1:500 dilution from a representative lot detected E2 of Chikungunya virus in C6/36 mosquito cell lysates either mock-infected (negative) or CHIKV-infected (positive). (Courtesy of Jody Peters, Ph.D., University of Queensland, Australia).
Neutralizing Analysis: A representative lot efficiently neutralized different CHIKV isolates in Neutralizing applications (Goh, L.Y., et. al. (2013). Clin Immunol. 149(3):487-97).
Western Blotting Analysis: A representative lot detected CHIKV E2 in Western Blotting applications (Goh, L.Y., et. al. (2013). Clin Immunol. 149(3):487-97).
ELISA Analysis: A representative lot detected CHIKV E2 in ELISA applications (Goh, L.Y., et. al. (2013). Clin Immunol. 149(3):487-97).
Neutralizing Analysis: A representative lot efficiently neutralized different CHIKV isolates in Neutralizing applications (Goh, L.Y., et. al. (2013). Clin Immunol. 149(3):487-97).
Western Blotting Analysis: A representative lot detected CHIKV E2 in Western Blotting applications (Goh, L.Y., et. al. (2013). Clin Immunol. 149(3):487-97).
ELISA Analysis: A representative lot detected CHIKV E2 in ELISA applications (Goh, L.Y., et. al. (2013). Clin Immunol. 149(3):487-97).
Biochem/physiol Actions
Clone 1.3A2 specifically detects E2 glycoprotein in Chikungunya virus. May recognize multiple epitopes on E2 protein.
Physical form
Purified mouse monoclonal antibody IgG2a in PBS without azide.
Preparation Note
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
Isotype testing: Identity Confirmation by Isotyping Test.
Isotyping Analysis: The identity of this monoclonal antibody is confirmed to be mouse IgG2a by isotype testing.
Isotyping Analysis: The identity of this monoclonal antibody is confirmed to be mouse IgG2a by isotype testing.
Other Notes
Chikungunya virus.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.