Skip to Content
Merck

MABD85

Anti-LIMD1 Antibody, clone 3F2/C6

clone 3F2/C6, from mouse

Synonym(s):

LIM domain-containing protein 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
3F2/C6, monoclonal
Application:
ICC, IHC, IP, WB
Citations:
3
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F2/C6, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LIMD1(8994)

General description

LIMD1 (LIM domains-containing protein 1) is a tumor suppressor and member of the Ajuba family of LIM domain-containing proteins. LIMD1 interacts with retinoblastoma protein (pRB), inhibits E2F-mediated transcription, and suppresses the expression of the majority of genes with E2F1-responsive elements. Recently, LIMD1 has been implicated in regulating cellular processes involved in hypoxia. LIMD1 acts as a molecular scaffold, simultaneously binding the prolyl hydroxylases (PHDs) and von Hippel Lindau tumor suppressors (VHL), thereby assembling a PHD-LIMD1-VHL protein complex and creating an enzymatic niche that enables efficient degradation of HIF-1α.
~72 kDa observed

Immunogen

Recombinant protein corresponding to human LIMD1

Application

Detect LIMD1 using this Anti-LIMD1 Antibody, clone 3F2/C6 validated for use in ICC, IHC, Immunoprecipitation, Western Blotting.
Research Category
Apoptosis & Cancer

Stem Cell Research
Research Sub Category
Oncoproteins

Oxidative Stress
This antibody has been shown to work in Immunocytochemical applications (Foxler, D., et al. (2012).
This antibody has been shown to work in Immunohistochemical applications (Sharp, T., et al. (2008).
This antibody has been shown to work in Immunoprecipitation applications (Courtesy of Dr. Tyson Sharp, Cancer Research, UK)

Biochem/physiol Actions

This antibody recognizes human LIMD1.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
H1299 cell lysates.
Evaluated by Western Blot in H1299 cell lysates.
Western Blot Analysis: A 1:2000 dilution of this antibody detected LIMD1 in H1299 cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library