Skip to Content
Merck
All Photos(1)

Key Documents

MAB6021

Sigma-Aldrich

Anti-Cytochrome P450 Aldosterone Synthase Antibody, clone 1G5

culture supernatant, clone 1G5, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
1G5, monoclonal
application:
ELISA
ICC
IHC
WB
species reactivity:
rat, mouse
technique(s):
ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable
citations:
5

biological source

mouse

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

1G5, monoclonal

species reactivity

rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Aldosterone synthase (or 18-hydroxylase) is a steroid hydroxylase cytochrome P450 oxidase enzyme involved in the generation of aldosterone. This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the mitochondrial inner membrane. The enzyme has steroid 18-hydroxylase activity to synthesize aldosterone and 18-oxocortisol as well as steroid 11 beta-hydroxylase activity. Mutations in this gene cause corticosterone methyl oxidase deficiency. (Pascoe, L, et al. (1992) "Mutations in the human CYP11B2 (aldosterone synthase) gene causing corticosterone methyloxidase II deficiency." PNAS, USA. 89:496-5000).

Specificity

Rat cytochrome p450 aldosterone synthase

Immunogen

Aldo-1-MAP (aa 175-192) (KVRQNARGSLTMDVQQSL-MAP)

Application

This Anti-Cytochrome P450 Aldosterone Synthase Antibody, clone 1G5 is validated for use in ELISA, IC, IH(P), WB for the detection of Cytochrome P450 Aldosterone Synthase.
Western blot: recognizes ~48kDa band in adrenal zona glomerulosa mitochondrial preparations. The enzyme is expressed best in adrenal zona glomerulosa tissues, expression in kidney is virtually undetectable.
Immunohistochemistry: 1:50-1:200 , ABC, enzymatic detection; clone works best with STF fixed tissues that are paraffin embedded. STF is available from Streck Laboratories, www.streck.com. Formalin fixation has been successfully reported but working dilutions are much lower (see Fujisawa, et al). Recommended counter stain is Gill′s Haematoxylin.
Immunocytochemistry: acetone fixed cells, 1:20-1:200.
ELISA
Optimal working dilutions must be determined by end user.

Physical form

Unpurified tissue culture supernatant containing 50% glycerol and 0.1% sodium azide as a preservative.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cherish Chong et al.
The Journal of endocrinology, 232(3), 525-534 (2017-01-18)
We posit the existence of a paracrine/autocrine negative feedback loop, mediated by the mineralocorticoid receptor (MR), regulating aldosterone secretion. To assess this hypothesis, we asked whether altering MR activity in zona glomerulosa (ZG) cells affects aldosterone production. To this end
Ana M Briones et al.
Hypertension (Dallas, Tex. : 1979), 59(5), 1069-1078 (2012-04-12)
We reported aldosterone as a novel adipocyte-derived factor that regulates vascular function. We aimed to investigate molecular mechanisms, signaling pathways, and functional significance of adipocyte-derived aldosterone and to test whether adipocyte-derived aldosterone is increased in diabetes mellitus-associated obesity, which contributes
Zheng Fan et al.
Cell reports, 43(7), 114395-114395 (2024-06-28)
Macrophages play crucial roles in organ-specific functions and homeostasis. In the adrenal gland, macrophages closely associate with sinusoidal capillaries in the aldosterone-producing zona glomerulosa. We demonstrate that macrophages preserve capillary specialization and modulate aldosterone secretion. Using macrophage-specific deletion of VEGF-A
Xiaoli Zhou et al.
Journal of the American Society of Nephrology : JASN, 27(6), 1765-1776 (2015-10-17)
Deficiency of the antiaging gene Klotho (KL) induces renal damage and hypertension through unknown mechanisms. In this study, we assessed whether KL regulates expression of CYP11B2, a key rate-limiting enzyme in aldosterone synthesis, in adrenal glands. We found that haplodeficiency
Arthur D Zimmerman et al.
Frontiers in endocrinology, 12, 697505-697505 (2021-08-03)
Atrazine is one of the most commonly used pre-emergence and early post-emergence herbicides in the world. We have shown previously that atrazine does not directly stimulate the pituitary or adrenal to trigger hormone release but acts centrally to activate a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service