Skip to Content
Merck
All Photos(1)

Key Documents

MAB4360A4

Sigma-Aldrich

Anti-TRA-1-60 Antibody, clone TRA-1-60, Alexa Fluor 488 Conjugate

clone TRA - 1-60, from mouse, ALEXA FLUOR 488

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
conjugate:
ALEXA FLUOR 488
application:
ICC
clone:
TRA - 1-60, monoclonal
species reactivity:
human
citations:
5
technique(s):
immunocytochemistry: suitable

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified antibody

antibody product type

primary antibodies

clone

TRA - 1-60, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable

isotype

IgM

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PODXL(5420)

General description

Human embryonal carcinoma (EC) cells are the stem cells of teratocarcinomas, and they are key components of germ cell tumors (GCTs). They express several high molecular weight glycoprotein antigens that are down-regulated upon differentiation. One of these antigens, defined by monoclonal antibody TRA-1-60, can be detected in the serum of GCT patients and provides a useful complement to the established serum markers human chorionic gonadotropin and α-fetoprotein, especially in those patients without elevated serum human chorionic gonadotropin or α-fetoprotein.

Specificity

This antibody reacts with TRA-1-60 antigen expressed upon the surface of human EC, EG, and ES cells. No immunoreactivity is seen with murine EC, EG or ES cells. TRA-1-81 (MAB4381A4) and TRA-1-60 monoclonal antibodies recognize antigens that are associated with a pericellular matrix proteoglycan.

Immunogen

Human embryonal carcinoma cell line 2102Ep

Application

Detect TRA-1-60 using this Anti-TRA-1-60 Antibody, clone TRA-1-60, Alexa Fluor 488 Conjugate validated for use in IC.
Research Category
Neuroscience

Quality

Evaluated by Immunocytochemistry in H9 human embryonic stem cells.

Immunocytochemistry Analysis: A 1:200 dilution of this antibody detected TRA-1-60 in H9 human embryonic stem cells.

Physical form

Purified mouse monoclonal IgM conjugated to Alexa Fluor 488 in PBS with 0.1% sodium azide and 15 mg/mL BSA.
size exclusion

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
H9 human embryonic stem cells

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yohei Hayashi et al.
Communications biology, 1, 218-218 (2018-12-12)
Conventional cell handling and sorting methods require manual labor, which decreases both cell quality and quantity. To purify adherent cultured cells, cell purification technologies that are high throughput without dissociation and can be utilized in an on-demand manner are expected.
Kung-Kai Kuo et al.
Stem cells (Dayton, Ohio), 34(11), 2613-2624 (2016-06-25)
The network of stemness genes and oncogenes in human patient-specific reprogrammed cancer stem cells (CSCs) remains elusive, especially in liver cancer. HepG2-derived induced pluripotent stem cell-like cells (HepG2-iPS-like cells) were generated by introducing Yamanaka factors and the knockdown vector shTP53.
Ning Sun et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(37), 15720-15725 (2009-10-07)
Ectopic expression of transcription factors can reprogram somatic cells to a pluripotent state. However, most of the studies used skin fibroblasts as the starting population for reprogramming, which usually take weeks for expansion from a single biopsy. We show here
Vanessa Sauer et al.
Cell transplantation, 25(12), 2221-2243 (2016-08-12)
Although several types of somatic cells have been reprogrammed into induced pluripotent stem cells (iPSCs) and then differentiated to hepatocyte-like cells (iHeps), the method for generating such cells from renal tubular epithelial cells shed in human urine and transplanting them
Zhongwen Li et al.
Cell death & disease, 10(10), 763-763 (2019-10-12)
Hepatocytes have been successfully generated from human pluripotent stem cells (hPSCs). However, the cost-effective and clinical-grade generation of hepatocytes from hPSCs still need to be improved. In this study, we reported the production of functional hepatocytes from clinical-grade human embryonic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service