Skip to Content
Merck

LS200

Hood Mounted Pipet Holder, Left Side Mount

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
eCl@ss:
32011202
Manufacturer/tradename:
Specialty Media
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Specialty Media

Quality Level

input

sample type cardiac stem cell(s)
sample type hematopoietic stem cell(s)
sample type epithelial cells
sample type: mouse embryonic stem cell(s)
sample type mesenchymal stem cell(s)
sample type: human embryonic stem cell(s)
sample type pancreatic stem cell(s)
sample type neural stem cell(s)
sample type induced pluripotent stem cell(s)

shipped in

ambient

storage temp.

15-25°C

General description

Hood Mounted Pipet Holder, Left Side Mount

Application

Research Category
Cell Culture

Packaging

each

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
LS20004053252587184