Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB269F

Sigma-Aldrich

Milli-Mark® Anti-Integrin β1 (CD29)-FITC conjugate Antibody, clone MB1.2

clone MB1.2, Milli-Mark®, from rat

Synonym(s):

CD29 antigen, Fibronectin receptor subunit beta, Integrin VLA-4 subunit beta, fibronectin receptor beta subunit, integrin VLA-4 beta subunit, integrin beta 1, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
conjugate:
FITC conjugate
application:
FACS
clone:
MB1.2, monoclonal
species reactivity:
mouse
citations:
1
technique(s):
flow cytometry: suitable

biological source

rat

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

MB1.2, monoclonal

species reactivity

mouse

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)

General description

Integrin beta 1, also known as CD29, is a 130 kDa transmembrane glycoprotein that forms noncovalent complexes with various Integrin alpha subunits (including alpha 1, alpha 2, alpha 3, alpha 4, alpha 5, and alpha 6, also known as CD49a, CD49b, CD49c, CD49d, CD49e, and CD49f, respectively) to form the functional receptors that bind to specific extracellular matrix proteins. Integrin receptors are involved in the regulation of a variety of important biological functions, including embryonic development, wound repair, hemostasis, and prevention of programmed cell death. They are also implicated in abnormal pathological states such as tumor directed angiogenesis, tumor cell growth, and metastasis. Interactions between integrins and the extracellular matrix lead to activation of signal transduction pathways and regulation of gene expression.

Specificity

Antibody recognizes integrin β1.

Application

Research Category
Cell Structure
Research Sub Category
Integrins
This Milli-Mark Anti-Integrin β1 (CD29)-FITC conjugate Antibody, clone MB1.2 is validated for use in FC for the detection of Integrin β1 (CD29) conjugate.

Quality

Evaluated by flow cytometry using NIH/3T3

Target description

88 kDa Calculated

Physical form

Protein A purified
Purified rat monoclonal IgG2a conjugated to FITC in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8°C protected from light for up to 6 months from date of receipt.

Analysis Note

Control
NIH/3T3 cells

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spatiotemporal control of phosphatidylinositol 4-phosphate by Sac2 regulates endocytic recycling.
Hsu, F; Hu, F; Mao, Y
The Journal of cell biology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service