Skip to Content
Merck
All Photos(1)

Key Documents

ECM330

Sigma-Aldrich

Human sE-Selectin-1 ELISA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

description

analytes available: sE-Selectin-1

species reactivity

human

technique(s)

ELISA: suitable

UniProt accession no.

General description

FUNCTION: SwissProt: P16581 # Expressed on cytokine induced endothelial cells and mediates their binding to leukocytes. The ligand recognized by ELAM-1 is sialyl-lewis X (alpha(1->3)fucosylated derivatives of polylactosamine that are found at the nonreducing termini of glycolipids).
SIZE: 610 amino acids; 66655 Da
SUBCELLULAR LOCATION: Membrane; Single-pass type I membrane protein.
SIMILARITY: SwissProt: P16581 ## Belongs to the selectin/LECAM family. & Contains 1 C-type lectin domain. & Contains 1 EGF-like domain. & Contains 6 Sushi (CCP/SCR) domains.

Components

· 1 aluminum pouches with a MicroWell Plate coated with Monoclonal Antibody (murine) to human E-Selectin · 1 vial (200 μL) HRP-Conjugated Anti-E-Selectin Monoclonal (murine) Antibody · 2 vials (100 ng/mL each, reconstituted) soluble E-selectin Standard, lyophilized {dissolve in 390 microliters for lot 0606032906}. · 1 vial lyophilized control serum · 1 bottle (50 mL) Wash Buffer Concentrate 20X (PBS with 1% TWEEN® 20)* · 1 vial (5 mL) Assay Buffer Concentrate 20X (PBS with 1% TWEEN® 20 and 10% BSA)* · 1 bottle (12 mL) Sample Diluent (buffered protein matrix)* · 1 vial (7 mL) Substrate Solution I (tetramethyl-benzidine) · 1 vial (7 mL) Substrate Solution II (0.02 % buffered hydrogen peroxide) · 1 vial (12 mL) Stop Solution (1M Phosphoric Acid) · 1 MicroWell Strip Holder · 2 Adhesive Plate Covers * reagents containing 0.01% thimerosal as preservative

Storage and Stability

Storage of Kit Components Store kit reagents between 2° and 8°C. Store the lyophilized control at -20°C. Immediately after use remaining reagents should be returned to cold storage (2° to 8°C) with the exception of reconstituted control (-20°C). Expiration date of the kit and reagents is stated on the kit box. Precautions: · Reagents are intended for research use only and are not for use in diagnostic or therapeutic procedures. · Do not mix or substitute reagents with those from other lots or other sources. · Do not use kit reagents beyond expiration date on label. · Do not expose kit reagents to strong light during storage or incubation. · Rubber or disposable latex gloves should be worn while handling kit reagents or specimens. · Some reagents contain thimerosal as preservative, which is highly toxic by inhalation, ingestion, or contact with skin. Thimerosal is a possible mutagen and should be handled accordingly. · Avoid contact of substrate solutions with oxidizing agents and metal. · In order to avoid microbial contamination or cross-contamination of reagents or specimens which may invalidate the test use disposable pipette tips and/or pipettes. · Use clean, dedicated reagent trays for dispensing the conjugate and substrate reagents. · Exposure to acids will inactivate the conjugate. · Glass-distilled water or deionized water must be used for reagent preparation. · Substrate solutions must be at room temperature prior to use.

Legal Information

MicroWell is a trademark of Thermo Fisher Scientific or its subsidiaries
TWEEN is a registered trademark of Croda International PLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service