Skip to Content
Merck
All Photos(2)

Key Documents

E5SU221

Millipore

NovaSeptum® GO Preloaded System with Autoclavable Bag, Vented, 5-port

sterile; β-irradiated

Synonym(s):

NovaSeptum® GO Autoclavable Bag, Vented

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104105

material

316 stainless steel cannula
polypropylene bag
silicone septum (platinum-cured)
silicone tubing

Quality Level

Agency

according to ISO 11137 (sterilization)
according to ISO 146441
meets USP 88 Biological Reactivity (all component materials)

sterility

sterile; β-irradiated

product line

NovaSeptum® GO

feature

autoclavable

parameter

-20-125 °C temp. range (-4-257 °F)
0.30 bar max. pressure (4.35 psi)

impurities

<2.15 EU/device bacterial endotoxins (LAL test)

fitting

male Luer-Lok® outlet (with female sterile vent filter)

application(s)

cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pharma/biopharma processes
sterile sampling
viral therapy

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Device configuration: single unit sampling - preloaded system
The NovaSeptum® GO pre-loaded disposable magazine is available with various standard configurations using NovaSeptum® GO high purity bags, autoclavable bags, bottles, transfer tubes or accurate volume (AV) syringe units. Other configurations are available upon request. For an optimal aseptic solution, use the NovaSeptum® GO pre-loaded disposable magazine together with a 1.5" or 2" NovAseptic® connector and NovaSeptum® GO pre-loaded starter kit (5/9 shot stainless steel base). Ideal for sampling from aseptic and sterile processes, the NovaSeptum® GO sterile sampling system provides consistently representative samples, safely and securely.

Application

Mobile Tanks
Autoclaved

Packaging

E5SU221-00215 (5x50 ml bags, 8 units per box)
E5SU221-00216 (5x100 ml bags, 8 units per box)
E5SU221-C0243 (5x250 ml bags, 7 units per box)
E5SU221-C0244 (5x1000 ml bags, 6 units per box)

Components

  • Septum: Platinum-cured silicone
  • Body: Polyester
  • Cannula: ASTM® 316 L Stainless steel
  • Fluid contact layer (bag): Polypropylene film
  • Tubing: Silicone
  • Outlet Tubing: Male Luer-Lok® with female sterile vent filter

Preparation Note

Units are integrity tested at regular intervals during manufacturing.

Aqueous extraction contains < 2.15 EU per device as determined using the Limulus Amebocyte Lysate (LAL) test.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
Luer-Lok is a registered trademark of Becton-Dickinson & Co.
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service