Skip to Content
Merck
All Photos(2)

Key Documents

ABS517

Sigma-Aldrich

Anti-PRMT8 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein arginine N-methyltransferase 8, Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 4, Protein arginine methyltransferase 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), porcine (based on 100% sequence homology), chicken (based on 100% sequence homology), baboon (based on 100% sequence homology), canine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRMT8(56341)

General description

Asymmetric dimethylation of arginine side chains in proteins is a frequent posttranslational modification, catalyzed by type I protein arginine methyltransferases (PRMTs). PRMT8 is a membrane-associated arginine methyltransferase that can both catalyze the formation of omega-N monomethylarginine (MMA) and asymmetrical dimethylarginine (aDMA). PRMT8 can form dimers with PRMT2 or PRMT1 and interacts with FYN kinase. PRMT8 is brain specific and it is a vertebrate restricted paralogue of PRMT1. In the past, arginine methylation was mainly observed on abundant proteins such as RNA-binding proteins and histones, but recent advances have revealed a plethora of arginine-methylated proteins implicated in a variety of cellular processes including signal transduction, epigenetic regulation and DNA repair pathways. PRMT8 has been shown to be critical for proper neuronal development and its expression is regionally restricted in the adult suggesting that PRMT8 is chiefly involved in the somatosensory and limbic systems, and a part of the motor nervous system development and maintenance.

Specificity

Other homologies: Homology: predicted to react with rat.

Immunogen

Epitope: C-terminal region of SAM-dependent methyltransferase PRMT-type domain
KLH-conjugated linear peptide corresponding to the SAM-dependent methyltransferase PRMT-type domain of Human PRMT8.

Application

Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected PRMT8 in human and mouse brain tissuse, and mouse thalamus.
Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Histones
This Anti-PRMT8 Antibody is validated for use in Western Blotting and Immunohistochemistry for the detection of PRMT8.

Quality

Evaluated by Western Blotting using human fetal brain tissue lysate.

Western Blotting Analysis: 0.1 µg/mL of this antibody detected PRMT8 in 10 µg of human fetal brain tissue lysate.

Target description

~48 kDa observed

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service