Skip to Content

ABS502

Anti-LTBP-1 Antibody

serum, from rabbit

Synonym(s):

Latent-transforming growth factor beta-binding protein 1, LTBP-1, Transforming growth factor beta-1-binding protein 1, TGF-beta1-BP-1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat

technique(s)

immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LTBP1(4052)

General description

Latent-transforming growth factor beta-binding protein 1 (LTBP-1) is also called Transforming growth factor beta-1-binding protein 1 (TGF-beta1-BP-1). A large latent complex of TGFB1 from platelets is composed of the TGFB1 molecule non-covalently associated with a disulfide-bonded complex of a dimer of the N-terminal propeptide of the TGFB1 precursor and LTBP1. LTBP1 does not bind directly to active TGFB1 only to proforms. LTBP1 appears to be involved in the secretion of TGFB1 to sites at which it is stored or activated, and it is critical in controlling the action of TGFB1. LTBP1 may also play a role in extracellular matrix building and calcium binding mediated signaling surrounding TGFB1 activated signaling pathways. Also, because of its critical role in TGFB1 metabolism, LTBP1 is an important player in many developmental processes including embryo implantation and growth. LTBP1 is also a player in at least two distinct breast cancer metastasis signatures and has both a long and short isoform that work to establish and maintain the spatial restriction of TGFbeta bioavailability during development of many tissues including mammary development.
~220 kDa observed. Uncharacterized band(s) may appear in some lysates.

Immunogen

His-tagged recombinant protein corresponding to human LTBP-1.

Application

Immunocytochemistry Analysis: A representative lot detected LTBP-1 in transfected HEK293 cells (Chen, Q., et al. (2007). JBC. 282(36):26418-26430).
Immunocytochemistry Analysis: A representative lot detected LTBP-1 in rat FRC cells (Chen, Q., et al. (2007). JBC. 282(36):26418-26430).
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Anti-LTBP-1 Antibody is validated for use in Western Blotting and Immunocytochemistry for the detection of LTBP-1.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in human lung tissue lysate using a non-reduced gel.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected LTBP-1 in 10 µg of human lung tissue lysate using a non-reduced gel.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library