Skip to Content
Merck
All Photos(2)

Key Documents

ABN174

Sigma-Aldrich

Anti-MNX1 (HB9) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Motor neuron and pancreas homeobox protein 1, Homeobox protein HB9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MNX1(3110)

General description

Motor neuron and pancreas homeobox protein 1 (MNX1) is a transcription factor that is predominantly expressed in lymphoid and pancreatic tissues. It is involved in the development and function of the pancreas. Abnormal expression of MNX1 results in an autosomal dominant condition known as Currarino syndrome.

Specificity

Other homologies: Zebrafish (94% sequence homology) and Xenopus (89% sequence homology).
This antibody recognizes a portion of the Homeobox DNA binding domain.

Immunogen

KLH-conjugated linear peptide corresponding to human MNX1.

Application

Anti-MNX1 (HB9) Antibody detects level of MNX1 (HB9) & has been published & validated for use in Western Blotting & IHC.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected MNX1 in human pancreas and mouse cerebral cortex tissues.
Research Category
Neuroscience
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blot in PANC-1 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected MNX1 in 10 µg of PANC-1 cell lysate.

Target description

~50 kDa observed. An uncharacterized band may appear at ~48 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
PANC-1 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gonzalo Perez-Siles et al.
Disease models & mechanisms, 13(2) (2020-01-24)
ATP7A encodes a copper-transporting P-type ATPase and is one of 23 genes in which mutations produce distal hereditary motor neuropathy (dHMN), a group of diseases characterized by length-dependent axonal degeneration of motor neurons. We have generated induced pluripotent stem cell
G Perez-Siles et al.
Scientific reports, 10(1), 9262-9262 (2020-06-07)
Charcot-Marie-Tooth (CMT) is a group of inherited diseases clinically and genetically heterogenous, characterised by length dependent degeneration of axons of the peripheral nervous system. A missense mutation (p.R158H) in the pyruvate dehydrogenase kinase 3 gene (PDK3) has been identified as
Daehwan Kim et al.
Translational neurodegeneration, 11(1), 16-16 (2022-03-12)
Amyotrophic lateral sclerosis (ALS) is characterized by a progressive loss of motor neurons (MNs), leading to paralysis, respiratory failure and death within 2-5 years of diagnosis. The exact mechanisms of sporadic ALS, which comprises 90% of all cases, remain unknown. In
Andrew Wang et al.
BMC endocrine disorders, 21(1), 185-185 (2021-09-15)
Cushing's disease (CD) is defined as hypercortisolemia caused by adrenocorticotropic hormone (ACTH)-secreting pituitary adenomas (corticotroph PA) that afflicts humans and dogs. In order to map common aberrant genomic features of CD between humans and dogs, we performed genomic sequencing and
Ilary Allodi et al.
Scientific reports, 6, 25960-25960 (2016-05-18)
The fatal disease amyotrophic lateral sclerosis (ALS) is characterized by the loss of somatic motor neurons leading to muscle wasting and paralysis. However, motor neurons in the oculomotor nucleus, controlling eye movement, are for unknown reasons spared. We found that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service