Skip to Content
Merck

Skip To

ABE1397

Anti-trimethyl STAT3 (Lys180) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Signal transducer and activator of transcription 3, Acute-phase response factor, trimethyl STAT3 (Lys180)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
mouse
Application:
IP, WB, inhibition assay
Citations:
1
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology), porcine (based on 100% sequence homology), orangutan (based on 100% sequence homology), chicken (based on 100% sequence homology), bovine (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable, inhibition assay: suitable (peptide), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys180 )

Gene Information

human ... STAT3(6774)

General description

Signal transducer and activator of transcription 3 (UniProt P40763; also known as Acute-phase response factor, DNA-binding protein APRF) is encoded by the STAT3 (also known as ADMIO, APRF, HIES) gene (Gene ID 6774) in human. STAT proteins are latent cytoplasmic transcription factors that have the dual function of signal transduction and activation of transcription. STATs are activated by tyrosine phosphorylation in response to different ligands, after which they translocate to the cell nucleus. The N-terminal region is highly homologous among the STAT proteins and surrounds a completely conserved arginine residue. STATs are a part of the JAK-STAT signaling pathway – a major pathway of the immune system. All cytokines transduce critical signals through this pathway. STAT3 is known to be activated by IFN alpha but not IFN beta. The transcription factors associated with STAT3 are cJun and cyclic AMP responsive enhancer binding protein (CREB). STAT3 gene knockout in mice is lethal at the early embryonic stage.
~88 kDa observed. Uncharacterized band(s) may appear in some lysates.

Immunogen

Epitope: STAT3 K180me3
KLH-conjugated linear peptide corresponding to the STAT3 K180me3 of human trimethyl STAT3 (Lys180)

Application

Immunoprecipitation Analysis: 5.0 µg from a representative lot immunoprecipitated STAT3 trimethylated on Lys180 in 0.5 mg of mouse embryonic stem cell lysate.
Peptide Inhibition Assay: Target specificity of a representative lot was confirmed by immunogen peptide blocking prior to Western blotting analysis (0.5 µg/mL) ) of mouse embryonic stem cell lysate (10 µg/lane) . Only the immunogen peptide, but not the corresponding peptide without the Lys180 trimethylation modification, blocked the detection of the target band.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-trimethyl STAT3 (Lys180) Antibody is validated for use in Western Blotting, Peptide Inhibition Assay, Immunoprecipitation for the detection of trimethyl STAT3 (Lys180).

Biochem/physiol Actions

Trimethylation site is present in all three human spliced isoforms. Reactivity toward Stat3 with mono- or dimethylated Lys180 has not been tested.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in mouse embryonic stem cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected STAT3 trimethylated on Lys180 in 10 µg of mouse embryonic stem cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4300034MABC1177ZRB1006
description

from rabbit, purified by affinity chromatography

description

affinity isolated antibody

description

clone 516 G10H9, from mouse

description

recombinant, expressed in HEK 293 cells

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit (recombinant)

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

purified antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

200

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable, inhibition assay: suitable (peptide)

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100, indirect immunofluorescence: 1:100-1:200, western blot: 1:500-1:1000

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:400-1:100, inhibition assay: 1:10,000 (peptide), western blot: suitable using 1:1,000

clone

polyclonal

clone

polyclonal

clone

516 G10H9, monoclonal

clone

6C3, recombinant monoclonal

species reactivity

mouse

species reactivity

mouse, human, rat

species reactivity

human

species reactivity

human


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters