Skip to Content
Merck

ABC463

Anti-Osteoprotegerin/TNFRSF11B Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Tumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TNFRSF11B(4982)

General description

Osteoprotegerin/TNFRSF11B, also known as Tumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, or Osteoprotegerin, (OPG), and encoded by the gene TNFRSF11B/OCIF/OPG, is a membrane bound receptor that acts as a decoy receptor for TNFSF11/RANKL and thereby helps to neutralize or regulate its function in osteoclastogenesis. Bone homeostasis is at least partially dependent upon the levels of TNFSF11 and TNFSF11B, and by acting as decoy or "extra" receptor for TNFSF11B the excess factor levels can be modulated. The activation of the receptors inhibits the activation of osteoclasts and promotes osteoclast apoptosis in vitro. OPG also seems to act as a decoy receptor for TNFSF10/TRAIL and help prevent against TRAIL mediated apoptosis, and TRAIL acts as antagonist for osteoclastogenesis, and thus again OPG appears to moderate osteoclast levels in a dynamic fashion. OPG is highly expressed in numerous tissues other than developing and adult bone germinal centers, but its exact role in these other tissues is still being examined.
~55 kDa observed. Protein is heavily glycosylated and often observed as a monomer at ~55-60 kDa (Cundy T., et al. (2002) Hum Mol Genet. 11(18):2119-27).

Immunogen

Epitope: TNFR-Cys 2 domain
KLH-conjugated linear peptide corresponding to the TNFR-Cys 2 domain of human Osteoprotegerin/TNFRSF11B.

Application

Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected Osteoprotegerin/TNFRSF11B in human kidney tissue.
This Anti-Osteoprotegerin/TNFRSF11B Antibody is validated for use in western blotting & IHC (Paraffin) for the detection of Osteoprotegerin/TNFRSF11B.

Biochem/physiol Actions

Immunogen is 92% identical in bovine and in mouse. Immunogen is 84% identical in rat.

Analysis Note

Evaluated by Western Blotting in MG63 conditioned media.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Osteoprotegerin/TNFRSF11B in 10 µg of MG63 conditioned media.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Sheng Bi et al.
Clinical and experimental dental research, 5(4), 377-388 (2020-01-17)
This study aimed to investigate the relationship between inflammation-related T-helper cell polarization and the receptor activator for nuclear factor-κB ligand (RANKL)/osteoprotegerin (OPG) ratio, which is associated with bone resorption or remodeling of chronic periodontitis patients. Gingival crevicular fluid (GCF) and

Global Trade Item Number

SKUGTIN
ABC46304053252977503

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service