Skip to Content
Merck

ABC304

Anti-Olfactomedin-4/OLFM4 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Olfactomedin-4, OLM4, Antiapoptotic protein GW112, G-CSF-stimulated clone 1 protein, hGC-1, hOLfD

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... OLFM4(10562)

General description

OLFM4, also known as Olfactomedin-4, Antiapoptotic protein GW112, G-CSF-stimulated clone 1 protein (hGC-1), or hOLfD, and encoded by the gene OLFM4/GW112 is a multimeric protein that has been found both extracellularly, interacting with cell surface lectins and cadherin proteins, and intracellularly within the mitochondria and cytoplasm. OLFM4 is expressed during myeloid lineage development but also in found in adult prostate, bone marrow, and stomach, as well as pancreatic cancer cells. The protein appears to govern cell proliferation in pancreatic cancer cells however in leukemic cell lines it inhibits cell growth and induces cell differentiation and apoptosis. The gene is induced by retinoic acid contact and signaling is linked via the NFkappaB pathway.
~64 kDa observed. The calculated molecular weight of this protein is 57 kDa, but can be observed at ~64 kDa due to high glycosylation.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human Olfactomedin-4/OLFM4.

Application

Anti-Olfactomedin-4/OLFM4 Antibody is an antibody against Olfactomedin-4/OLFM4 for use in western blotting & IHC.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: 2.0 µg/mL of this antibody detected Olfactomedin-4/OLFM4 in 10 µg of human liver tissue lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Olfactomedin-4/OLFM4 in human pancreatic cancer sections and human prostate tissue.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in human small intestine tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Olfactomedin-4/OLFM4 in 10 µg of human small intestine tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library