Skip to Content
Merck
All Photos(1)

Documents

AB3792

Sigma-Aldrich

Anti-V5 Epitope Tag Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-V5 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Recognizes V5 (GKPIPNPLLGLDST).

Immunogen

GKPIPNPLLGLDST (V5)

Application

Anti-V5 Epitope Tag Antibody is an antibody against V5 Epitope Tag for use in IC, IP & WB.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
Western blot

Immunocytochemistry

Immunoprecipitation

Optimal working dilutions must be determined by the end user.

Physical form

ImmunoAffinity Purified
Liquid in PBS, pH 7.2 with 0.09% sodium azide.

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Proteins tagged with V5

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Itsushi Minoura et al.
Nature communications, 7, 10058-10058 (2016-01-19)
Mutations in human β3-tubulin (TUBB3) cause an ocular motility disorder termed congenital fibrosis of the extraocular muscles type 3 (CFEOM3). In CFEOM3, the oculomotor nervous system develops abnormally due to impaired axon guidance and maintenance; however, the underlying mechanism linking
Role of lysyl oxidase propeptide in secretion and enzyme activity.
Grimsby, JL; Lucero, HA; Trackman, PC; Ravid, K; Kagan, HM
Journal of Cellular Biochemistry null
Rajesh Kumar Gandhirajan et al.
The Journal of biological chemistry, 291(22), 11596-11607 (2016-04-07)
Transcriptional co-activator with PDZ-binding motif (TAZ) and Yes-associated protein (YAP) are critical transcriptional co-activators downstream of the Hippo pathway involved in the regulation of organ size, tissue regeneration, proliferation, and apoptosis. Recent studies suggested common and distinct functions of TAZ
Sherry T Shu et al.
The Journal of biological chemistry, 292(7), 2670-2678 (2016-12-30)
The HIV-1 Nef accessory factor enhances viral infectivity, immune evasion, and AIDS progression. Nef triggers rapid down-regulation of CD4 via the endocytic adaptor protein 2 (AP-2) complex, a process linked to enhanced viral infectivity and immune escape. Here, we describe
Eleni Petsalaki et al.
The Journal of cell biology, 195(3), 449-466 (2011-10-26)
Aurora B kinase activity is required for successful cell division. In this paper, we show that Aurora B is phosphorylated at serine 331 (Ser331) during mitosis and that phosphorylated Aurora B localizes to kinetochores in prometaphase cells. Chk1 kinase is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service