Skip to Content
Merck
All Photos(1)

Key Documents

AB3036

Sigma-Aldrich

Anti-Uncoupling Protein 1 Antibody

Chemicon®, from rabbit

Synonym(s):

UCP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Ucp1(22227)

Specificity

Uncoupling Protein 1 (UCP1), mouse. Mouse/rat UCP1 is a 307 amino acid mitochondrial uncoupling protein (Bouillaud et al. 1986; Ridley et al. 1986; Kozak et al. 1988; Cassard et al. 1990; Miroux et al. 1993). It is only 59% homologous with UCP2 (Fleury et al. 1997). Human UCP1 is located on chromosome 4 (Bouillaud et al. 1986; Ridley et al. 1986; Kozak et al. 1988; Cassard et al. 1990; Miroux et al. 1993). UCP1 is primarily expressed in brown adipose tissues.

The antibody has not yet been tested for cross-reactivity to other species. The sequence used for immunization shows 100% homology to rat, 91% with golden hamster, 83% with rabbit and 75% with human and canine UCP1.

Immunogen

A 12 aa peptide at the C-terminus of the mouse/rat UCP1 (Bouillaud et al. 1986; Ridley et al. 1986; Kozak et al. 1988; Cassard et al. 1990; Miroux et al. 1993).

Application

Detect Uncoupling Protein 1 using this Anti-Uncoupling Protein 1 Antibody validated for use in ELISA & WB.
Western Blotting: 1-10 μg/mL using Chemiluminescence technique. An antibody made to this epitope has detected a protein of ~32 kDa in brown adipose tissues (Feil & Rafael 1994).

ELISA: 1:10,000-1:100,000; using 50-100 ng UCP1 control peptide/well.

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS with less than 0.1% sodium azide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Luo et al.
Frontiers in endocrinology, 13, 877152-877152 (2022-05-21)
Thermogenic adipocytes possess a promising approach to combat obesity with its capability promoting energy metabolism. We previously discovered that deletion of GPR30 (GPRKO), a presumably membrane-associated estrogen receptor, protected female mice from developing obesity, glucose intolerance, and insulin resistance when
Andrea V Rozo et al.
Molecular metabolism, 6(7), 748-759 (2017-07-14)
Adult obesity risk is influenced by alterations to fetal and neonatal environments. Modifying neonatal gut or neurohormone signaling pathways can have negative metabolic consequences in adulthood. Here we characterize the effect of neonatal activation of glucagon like peptide-1 (GLP-1) receptor
Melanie Lacaria et al.
PLoS genetics, 8(5), e1002713-e1002713 (2012-06-02)
The functional contribution of CNV to human biology and disease pathophysiology has undergone limited exploration. Recent observations in humans indicate a tentative link between CNV and weight regulation. Smith-Magenis syndrome (SMS), manifesting obesity and hypercholesterolemia, results from a deletion CNV

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service