Skip to Content

A012-M

NovaSeptum®Multi-sampling Unit Bag Rack

For use with 9-port holder, TC 64 for NAC-USI, NAC-USM

Synonym(s):

manifold bag rack, multi-sample bag rack, sterile sample bag rack

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
36101790
UNSPSC Code:
41104105
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

stainless steel (Manifold holder: Polyacetale copolymere)

sterility

non-sterile

product line

NovaSeptum®

packaging

pkg of 1 unit

L × W × H

300 mm (11.8 in.) × 60 mm (2.3 in.) × 290-490 mm (11.4-19.3 in.)

weight

0.5 kg

application(s)

sterile sampling

compatibility

for use with 9-port holder, TC 64 for NAC-USI, NAC-USM

General description

The NovaSeptum®Multi-Sampling Bag Rack is a fully adjustable stainless steel support accessory designed to hold and organize NovaSeptum®sampling units during aseptic fluid sampling operations. The rack is engineered for compatibility with 9-port TC 64 (Tri-Clamp 64 mm) holders, compatible with NAC-USI and NAC-USM (NovAseptic Connector) connectors. It includes a polyacetal copolymer manifold holder to cradle and stabilize multi-container manifold assemblies, supporting high-throughput sterile sampling from a single process connection point.

Application

  • The NovaSeptum® Bag Rack acts as a structured mechanical support frame that holds and organizes NovaSeptum® sampling units during aseptic fluid sampling operations.
  • It is particularly suited for use in biopharmaceutical manufacturing and cleanroom environments where controlled, contamination-free fluid sampling is required.

Features and Benefits

  • Fully adjustable to accommodate different process setups
  • The maximum load capacity of the rack goes up to 5 kgs
  • The structural components are manufactured from stainless steel in compliance with ASTM®316, while the screws and locking nuts are made from A4-grade stainless steel
  • A certificate of quality is included in each package for traceability and compliance
  • Compatible with bag, bottle and conical tube sampling units

Packaging

A complete bag rack unit, including all components, is packaged unassembled in a cardboard box and assigned a part number.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library