Skip to Content
Merck

8.55125

NovaPEG Rink Amide resin LL

Novabiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352111
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

NovaPEG, Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

General description

An excellent support for the synthesis of peptide amides by Fmoc SPPS. The support has excellent swelling properties in a wide range of solvents, including water, MeCN, MeOH, DCM, DMF, THF, and toluene. In comparative studies this resin was found to give better results than polystyrene-based supports in the synthesis of hydrophobic peptides. The high hydrophilicity was also found to benefit on-resin immunoassays with one-bead-one-peptide libraries [1].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.15 - 0.25 mmol/g
TNBS-Test: passes test
Swelling Volume (in CH₂Cl₂):
PEG based resin (35 - 100 mesh)

Other Notes

Replaces: 01-64-0483

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_c

Not applicable

flash_point_f

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library