Skip to Content
Merck
All Photos(1)

Key Documents

8.55051

Sigma-Aldrich

Biotin NovaTag® resin

Novabiochem®

Synonym(s):

N-Biotin-NÆ-Fmoc-ethylenediamine MPB-AM resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352209

product line

NovaSyn® TG
Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

15-25°C

General description

Biotin NovaTag resin is an ideal tool for the synthesis of peptides labeled with biotin. As the biotin moiety is incorporated into the linker, its use eliminates the problems associated with biotin-labeling of peptides, namely poor solubility and sluggish coupling kinetics of biotin and its derivatives. Following peptide assembly under standard Fmoc SPPS conditions, cleavage with TFA affords the peptide labeled with biotin. For a recent applications see [1,2].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] M. Carraz, et al. (2009) Chemistry & Biology, 16, 702.
[2] S. Millward, et al. (2007) Chem. Biol., 2, 625.

Linkage

Replaces: 04-12-3901

Analysis Note

Color (visual): white to light yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.35 - 0.65 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer metrix is copoly (styrene - 1% DVB), 100 - 200 mesh.

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVATAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Biotin-labelled peptides find applications in immunology and histochemistry for affinity purification and receptor localization.

Biotin-labelled peptides find applications in immunology and histochemistry for affinity purification and receptor localization.

Biotin-labelled peptides find applications in immunology and histochemistry for affinity purification and receptor localization.

Biotin-labelled peptides find applications in immunology and histochemistry for affinity purification and receptor localization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service