Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C39H47N3O9S
Molecular Weight:
733.87
NACRES:
NA.22
UNSPSC Code:
12352209
Quality Segment
product line
Novabiochem®
form
powder
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
thiol
storage temp.
15-25°C
General description
A cysteine-based pseudoproline dipeptide. It prevents aggregation during Fmoc SPPS and enhances product purity in an identical manner to standard pseuproline dipeptides. The pseudoproline ring is cleaved with TFA, regenerating the native Cys residue.
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Overcoming Aggregation in Fmoc SPPS
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Overcoming Aggregation in Fmoc SPPS
Analysis Note
Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity Cysteine: ≥ 99.5 % (a/a)
Enantiomeric purity second amino acid: ≥ 99.0 % (a/a)
Purity (TLC(157A)): ≥ 97 %
Purity (TLC(CMA2)): ≥ 97 %
Assay (HPLC, area%): ≥ 97.0 % (a/a)
Ethyl acetate (HS-GC): ≤ 0.50 %
Acetate (IC): ≤ 0.05
To see the solvent systems used for TLC of Novabiochem® products please click here.
The product is a racemic mixture.
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity Cysteine: ≥ 99.5 % (a/a)
Enantiomeric purity second amino acid: ≥ 99.0 % (a/a)
Purity (TLC(157A)): ≥ 97 %
Purity (TLC(CMA2)): ≥ 97 %
Assay (HPLC, area%): ≥ 97.0 % (a/a)
Ethyl acetate (HS-GC): ≤ 0.50 %
Acetate (IC): ≤ 0.05
To see the solvent systems used for TLC of Novabiochem® products please click here.
The product is a racemic mixture.
Legal Information
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.