Skip to Content

8.52151

Fmoc-D-Tyr(tBu)-OH

≥98.0% (HPLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-D-Tyr(tBu)-OH, N-α-Fmoc-O-t.-butyl-D-tyrosine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C28H29NO5
CAS Number:
Molecular Weight:
459.53
UNSPSC Code:
12352209
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fmoc-D-Tyr(tBu)-OH, Novabiochem®

Quality Segment

product line

Novabiochem®

assay

≥97.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

hydroxyl

storage temp.

2-30°C

SMILES string

N([C@H](Cc4ccc(cc4)OC(C)(C)C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C28H29NO5/c1-28(2,3)34-19-14-12-18(13-15-19)16-25(26(30)31)29-27(32)33-17-24-22-10-6-4-8-20(22)21-9-5-7-11-23(21)24/h4-15,24-25H,16-17H2,1-3H3,(H,29,32)(H,30,31)/t25-/m1/s1

InChI key

JAUKCFULLJFBFN-RUZDIDTESA-N

General description

Standard building block of introduction of D-tyrosine amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 97.0 %
Water (K. F.): ≤ 1.00 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-13-1037

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Fmoc-D-Tyr(tBu)-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library