Skip to Content
Merck
All Photos(1)

Key Documents

8.52070

Sigma-Aldrich

Fmoc-Thr(PO(OBzl)OH)-OH

Novabiochem®

Synonym(s):

Fmoc-Thr(PO(OBzl)OH)-OH, N-α-Fmoc-O-benzyl-L-phosphothreonine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26NO8P
CAS Number:
Molecular Weight:
511.46
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥90.0% (acidimetric)
≥98% (TLC)
≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

15-25°C

SMILES string

[P](=O)(O[C@H]([C@@H](NC(=O)OCC2c3c(cccc3)c4c2cccc4)C(=O)O)C)(OCc1ccccc1)O

InChI

1S/C26H26NO8P/c1-17(35-36(31,32)34-15-18-9-3-2-4-10-18)24(25(28)29)27-26(30)33-16-23-21-13-7-5-11-19(21)20-12-6-8-14-22(20)23/h2-14,17,23-24H,15-16H2,1H3,(H,27,30)(H,28,29)(H,31,32)/t17-,24+/m0/s1

InChI key

HOFDVXHILSPFNS-BXKMTCNYSA-N

General description

An excellent building block for the preparation of phosphothreonine-containing peptides [1,2] by Fmoc SPPS. This derivative can be introduced using standard activation methods, such as PyBOP® and TBTU. The monoprotected phosphothreonine residue once incorporated is stable to piperidine. Using this reagent, even peptides containing multiple phosphorylation sites can be prepared efficiently by standard Fmoc SPPS methods [1].A paper describes the use of this derivative in the preparation of phospholamban [3], a 52 residue peptide containing both phosphoserine and phosphothreonine.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] P. White & J. Beythien in ′Innovations & Perspectives in Solid Phase Synthesis and Combinatorial Libraries, 4th International Symposium′, Mayflower Scientific Ltd., Birmingham, 1996, pp. 557.
[2] T. Vorherr, et al. (1995) Bioorg. Med. Chem. Lett., 5, 2661.
[3] H. Schmid, et al., Poster 423 presented at the 15th American Peptide Symposium, Nashville, 1997.

Application


  • Development of highly selective 1, 2, 3-triazole-containing peptidic Polo-like kinase 1 Polo-box domain-binding inhibitors: This study explores the synthesis of inhibitors using Fmoc-Thr(PO(OBzl)OH)-OH among other amino acids, showcasing its utility in developing targeted cancer therapies (Zhao et al., 2019).



Linkage

Replaces: 04-12-1155

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in DMF): -6.5 - -3.5 °
Purity (TLC(CMA1)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 90.0 %
Water (K. F.): ≤ 2.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.1 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
PyBOP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service