Skip to Content
Merck
All Photos(1)

Key Documents

8.20788

Sigma-Aldrich

4-Methylcyclohexanone

for synthesis

Synonym(s):

4-Methylcyclohexanone, Tetrahydro-p-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(CH3)C6H9(=O)
CAS Number:
Molecular Weight:
112.17
MDL number:
UNSPSC Code:
12352115
EC Index Number:
209-665-3
NACRES:
NA.22

vapor pressure

20 hPa ( 65 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

800 mg/kg LD50, oral (Rat)

bp

169-171 °C/1013 hPa

mp

-41 °C

transition temp

flash point 48 °C

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C1CCC(CC1)C

InChI

1S/C7H12O/c1-6-2-4-7(8)5-3-6/h6H,2-5H2,1H3

InChI key

VGVHNLRUAMRIEW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.915 - 0.917
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service