Skip to Content
Merck

8.20744

Potassium-O-ethyldithiocarbonate

for synthesis

Synonym(s):

Potassium-O-ethyldithiocarbonate, Potassium ethylxanthogenate, Potassium xanthogenate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3H5KOS2
CAS Number:
Molecular Weight:
160.30
UNSPSC Code:
12352107
EC Index Number:
205-439-3
NACRES:
NA.22
MDL number:
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

potency

1700 mg/kg LD50, oral (Rat)

pH

9-10 (20 °C, 100 g/L in H2O)

mp

209-214 °C (decomposes)

solubility

1090 g/L

bulk density

400‑600 kg/m3

storage temp.

2-30°C

SMILES string

[K+].[S-]C(=S)OCC

InChI

1S/C3H6OS2.K/c1-2-4-3(5)6;/h2H2,1H3,(H,5,6);/q;+1/p-1

InChI key

JCBJVAJGLKENNC-UHFFFAOYSA-M

Analysis Note

Assay (iodometric): ≥ 97.0 %
Identity (IR): passes test


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk

WGK 3

flash_point_f

204.8 °F

flash_point_c

96 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library