Skip to Content
Merck
All Photos(1)

Key Documents

8.18203

Sigma-Aldrich

(9-Fluorenylmethyl) chloroformate

for synthesis

Synonym(s):

(9-Fluorenylmethyl) chloroformate, Chloroformic acid 9-fluorenylmethyl ester, FMOCCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11ClO2
CAS Number:
Molecular Weight:
258.70
MDL number:
UNSPSC Code:
12352108
EC Index Number:
249-313-6
NACRES:
NA.22
grade:
for synthesis
application(s):
peptide synthesis

grade

for synthesis

Quality Level

form

powder

dilution

(for synthesis)

mp

60-63 °C

application(s)

peptide synthesis

storage temp.

15-25°C

SMILES string

ClC(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C15H11ClO2/c16-15(17)18-9-14-12-7-3-1-5-10(12)11-6-2-4-8-13(11)14/h1-8,14H,9H2

InChI key

IRXSLJNXXZKURP-UHFFFAOYSA-N

General description

(9-Fluorenylmethyl) chloroformate, also known as Fmoc chloride, is a highly versatile reagent with varied applications in organic synthesis. It is most frequently used to introduce the base-labile Fmoc-protecting group to amine functionalities, particularly during the production of Fmoc-protected amino acids. Fmoc-chloride has also been used to generate mixed carboxylic and carbonic anhydrides to facilitate amide and ester bond formation. It is soluble in common organic solvents such as dichloromethane, tetrahydrofuran, and dimethylformamide.

Application

Recent applications of (9-Fluorenylmethyl) chloroformate include:
  • (9-Fluorenylmethyl) chloroformate can be used as a coupling reagent.
  • In the synthesis of amino acid esters. Fmoc chloride activates the carboxylic acid group of the amino acid, allowing it to react with an alcohol to form the ester.
  • In the preparation of lysogangliosides.
  • In the solid-phase peptide synthesis of DOPA-containing peptides.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Reactive Amino Acid Esters by Using 9-Fluorenylmethyl Chloroformate
Yamamoto H and Wu A
Synfacts, 16, 1247-1247 (2020)
9-Fluorenylmethoxycarbonyl amino-protecting group
Carpino LA and Han GY
The Journal of Organic Chemistry, 37, 3404-3404 (1972)
Synthesis of peptides containing DOPA (3, 4-dihydroxyphenylalanine)
Sever MJ and Wilker JJ
Tetrahedron, 6139-6146 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service