Skip to Content
Merck
All Photos(1)

Documents

8.17068

SAFC

Sodium formaldehydesulfoxylate hydrate

stabilized with sodium carbonate

Synonym(s):

Sodium formaldehydesulfoxylate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3NaO3S · xH2O (x ca. 2)
CAS Number:
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-739-4

vapor pressure

2.68 hPa ( 20 °C)

Quality Level

form

solid

pH

9.5-10.5 (20 °C, 100 g/L in H2O)

mp

63 °C

density

1.8 g/cm3 at 20 °C

application(s)

pharma/biopharma processes

storage temp.

2-30°C

InChI

1S/CH3O3S.Na.2H2O/c2-1-5(3)4;;;/h1H2,(H,3,4);;2*1H2/q-1;+1;;

InChI key

VJWVWUHJLZEHOU-UHFFFAOYSA-N

Application

Sodium formaldehydesulfoxylate hydrate is typically used as a preservative, antioxidant and stabilizer.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 2

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service